(4S,4'S)-1,1',4,4'-Tetraisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%

Supplier: Ambeed
Warning

A1501177-250MG A1501177-1G
77624-680EA 272.95 USD
77624-680 77875-730
(4S,4'S)-1,1',4,4'-Tetraisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-1,1',4,4'-Tetraisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole

Precaución: for Research Use Only

Formula: C₁₈H₃₄N₄
MDL Number: MFCD34598985
CAS Number: 2374958-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR