(4S,4'S)-1,1'-Di([1,1'-biphenyl]-4-yl)-4,4'-diisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%

Supplier: Ambeed
Warning

A1501176-100MG A1501176-1G A1501176-250MG
77624-674EA 145.23 USD
77624-674 77624-676 77624-678
(4S,4'S)-1,1'-Di([1,1'-biphenyl]-4-yl)-4,4'-diisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-1,1'-Di([1,1'-biphenyl]-4-yl)-4,4'-diisopropyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole

Precaución: for Research Use Only

Formula: C₃₆H₃₈N₄
MW: 526.71 g/mol
CAS Number: 2374958-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR