(4S,4'S)-4,4'-Diisopropyl-1,1'-bis(4-methoxyphenyl)-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
Supplier: Ambeed
|
Warning
|
A1501174-100MG
A1501174-250MG
77597-766EA
154.78
USD
77597-766
77624-670
(4S,4'S)-4,4'-Diisopropyl-1,1'-bis(4-methoxyphenyl)-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-4,4'-Diisopropyl-1,1'-bis(4-methoxyphenyl)-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole
Precaución: for Research Use Only
Formula:
C₂₆H₃₄N₄O₂ MW: 434.57 g/mol |
CAS Number:
2374958-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...