(4S,4'S)-4,4'-Diisopropyl-1,1'-bis(3-isopropylphenyl)-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%

Supplier: Ambeed
Warning

A1501172-100MG A1501172-250MG
77597-762EA 132.23 USD
77597-762 77624-666
(4S,4'S)-4,4'-Diisopropyl-1,1'-bis(3-isopropylphenyl)-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-4,4'-Diisopropyl-1,1'-bis(3-isopropylphenyl)-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole

Precaución: for Research Use Only

Formula: C₃₀H₄₂N₄
MW: 458.681 g/mol
MDL Number: MFCD34598728
CAS Number: 2374958-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR