(4S,4'S)-4,4'-Diisopropyl-1,1'-di-p-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%

Supplier: Ambeed
Warning

A1501170-100MG A1501170-250MG A1501170-1G
77597-758EA 112.75 USD
77597-758 77597-760 77624-664
(4S,4'S)-4,4'-Diisopropyl-1,1'-di-p-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole ≥97%
(4S,4'S)-4,4'-Diisopropyl-1,1'-di-p-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole

Precaución: for Research Use Only

Formula: C₂₆H₃₄N₄
MW: 402.5750 g/mol
CAS Number: 1827669-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR