5-Bromo-1,10-phenanthroline hydrate ≥95%

Supplier: Ambeed
Warning

A150072-1G A150072-250MG A150072-5G
77615-214EA 31.93 USD
77615-214 77746-042 77746-043
5-Bromo-1,10-phenanthroline hydrate ≥95%
5-Bromo-1,10-phenanthroline hydrate

Precaución: for Research Use Only

Formula: C₁₂H₉BrN₂O
MDL Number: MFCD28384130
CAS Number: 855360-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR