Bis(4-(diphenylamino)phenyl)methanone ≥95%

Supplier: Ambeed
Warning

A1500276-1G A1500276-100MG A1500276-250MG
77624-650EA 106.09 USD
77624-650 77875-705 77875-706
Bis(4-(diphenylamino)phenyl)methanone ≥95%
Bis(4-(diphenylamino)phenyl)methanone

Precaución: for Research Use Only

Formula: C₃₇H₂₈N₂O
MDL Number: MFCD01851488
CAS Number: 2873-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR