4-((4-Aminobutyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide-C4-NH2 HCl

A1499958-100MG A1499958-250MG A1499958-50MG
77597-754EA 152.73 USD
77597-754 77624-642 77624-644
4-((4-Aminobutyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
4-((4-Aminobutyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₄O₄
MW: 380.83 g/mol
MDL Number: MFCD31730832
CAS Number: 2162120-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR