(2S,4R)-1-((S)-2-(2-Aminoacetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC-C1-NH2 HCl

A1499955-50MG A1499955-100MG A1499955-1G A1499955-250MG
77624-640EA 181.28 USD
77624-640 77624-634 77624-636 77624-638
(2S,4R)-1-((S)-2-(2-Aminoacetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride ≥95%
(2S,4R)-1-((S)-2-(2-Aminoacetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₃₄ClN₅O₄S
MW: 524.08 g/mol
MDL Number: MFCD32903573
CAS Number: 2361120-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR