2-Bromo-N-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)acetamide ≥98%

Supplier: Ambeed
Danger

Synonyms: Bromoacetamide-PEG3-propargyl

A1499839-250MG A1499839-100MG A1499839-1G
77597-752EA 269.58 USD
77597-752 77624-630 77624-632
2-Bromo-N-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)acetamide ≥98%
2-Bromo-N-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)acetamide

Precaución: for Research Use Only

Formula: C₁₁H₁₈BrNO₄
MDL Number: MFCD32642009
CAS Number: 932741-11-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR