3-(2-(3-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)ethoxy)propanoic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S,R,S)-AHPC-PEG2-acid
A1499786-100MG
A1499786-250MG
77624-626EA
257.5
USD
77624-626
77624-628
3-(2-(3-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)ethoxy)propanoic acid ≥95%
3-(2-(3-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)ethoxy)propanoic acid
Precaución: for Research Use Only
Formula:
C₃₀H₄₂N₄O₈S MW: 618.74 g/mol |
MDL Number:
MFCD32670313 CAS Number: 2172820-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...