tert-Butyl (2-(2-(2-oxoethoxy)ethoxy)ethyl)carbamate ≥95%

Supplier: Ambeed
Warning

A1499337-1G A1499337-250MG
77875-693EA 941 USD
77875-693 77875-694
tert-Butyl (2-(2-(2-oxoethoxy)ethoxy)ethyl)carbamate ≥95%
tert-Butyl (2-(2-(2-oxoethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₅
MDL Number: MFCD31647292
CAS Number: 1229024-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR