3-Morpholinobicyclo[1.1.1]pentan-1-amine dihydrochloride ≥98%

Supplier: Ambeed
Warning

A1497955-100MG A1497955-1G A1497955-250MG
77875-687EA 460 USD
77875-687 77875-688 77875-689
3-Morpholinobicyclo[1.1.1]pentan-1-amine dihydrochloride ≥98%
3-Morpholinobicyclo[1.1.1]pentan-1-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈Cl₂N₂O
MDL Number: MFCD29060072
CAS Number: 2514758-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR