3,3-Dimethyl-2,3-dihydropyrazolo[5,1-b]oxazole-7-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A1497564-250MG A1497564-100MG
77592-822EA 414.1 USD
77592-822 77624-614
3,3-Dimethyl-2,3-dihydropyrazolo[5,1-b]oxazole-7-carboxylic acid ≥97%
3,3-Dimethyl-2,3-dihydropyrazolo[5,1-b]oxazole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MW: 182.17 g/mol
CAS Number: 2676863-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR