tert-Butyl (3-bromocyclobutyl)carbamate ≥97%

Supplier: Ambeed
Warning

A1496991-100MG A1496991-1G A1496991-250MG
77624-602EA 60.77 USD
77624-602 77875-662 77875-663
tert-Butyl (3-bromocyclobutyl)carbamate ≥97%
tert-Butyl (3-bromocyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₆BrNO₂
MDL Number: MFCD31691123
CAS Number: 1936399-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR