4-((7-Aminoheptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione ≥95%

Supplier: Ambeed
Warning

A1496746-100MG A1496746-50MG
77875-660EA 384 USD
77875-660 77875-661
4-((7-Aminoheptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione ≥95%
4-((7-Aminoheptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₂₀H₂₆N₄O₄
MDL Number: MFCD31730835
CAS Number: 2093387-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR