2-(Pyrrolidin-1-yl)pyrimidine-4-carboxylic acid hydrochloride ≥95%

Supplier: Ambeed
Warning

A1496655-100MG A1496655-250MG
77624-598EA 53.56 USD
77624-598 77875-659
2-(Pyrrolidin-1-yl)pyrimidine-4-carboxylic acid hydrochloride ≥95%
2-(Pyrrolidin-1-yl)pyrimidine-4-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClN₃O₂
MDL Number: MFCD32066591
CAS Number: 2361643-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR