4-(4-(1-Isopropyl-7-oxo-1,4,6,7-tetrahydrospiro[indazole-5,4'-piperidine]-1'-carbonyl)-6-methoxypyridin-2-yl)benzoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: PF-05221304

A1496582-10MG A1496582-1MG A1496582-5MG
77597-738EA 82 USD
77597-738 77597-740 77597-742
4-(4-(1-Isopropyl-7-oxo-1,4,6,7-tetrahydrospiro[indazole-5,4'-piperidine]-1'-carbonyl)-6-methoxypyridin-2-yl)benzoic acid ≥95%
4-(4-(1-Isopropyl-7-oxo-1,4,6,7-tetrahydrospiro[indazole-5,4'-piperidine]-1'-carbonyl)-6-methoxypyridin-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₄O₅
MW: 502.56 g/mol
MDL Number: MFCD33023235
CAS Number: 1370448-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR