5-Bromo-2-chloropyridine-4-carbaldehyde ≥97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-chloro-pyridine-4-carbaldehyde, 5-Bromo-2-chloroisonicotinaldehyde

A1496402-1G A1496402-5G
77592-820EA 14.35 USD
77592-820 77624-590
5-Bromo-2-chloropyridine-4-carbaldehyde ≥97%
5-Bromo-2-chloropyridine-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
MW: 220.46 g/mol
MDL Number: MFCD11847346
CAS Number: 1060802-23-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR