5-Chlorobenzo[d][1,2,3]thiadiazole 95+%

Supplier: Ambeed
Warning

A1496069-1G A1496069-100MG A1496069-250MG
77624-578EA 120.51 USD
77624-578 77875-637 77875-638
5-Chlorobenzo[d][1,2,3]thiadiazole 95+%
5-Chlorobenzo[d][1,2,3]thiadiazole

Precaución: for Research Use Only

Formula: C₆H₃ClN₂S
MDL Number: MFCD18449415
CAS Number: 29241-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR