5,7-Di-tert-butyl-3-(2,6-diisopropylphenyl)benzo[d]oxazol-3-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Danger

A1495852-250MG A1495852-100MG
77597-724EA 124.03 USD
77597-724 77624-574
5,7-Di-tert-butyl-3-(2,6-diisopropylphenyl)benzo[d]oxazol-3-ium tetrafluoroborate ≥97%
5,7-Di-tert-butyl-3-(2,6-diisopropylphenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₇H₃₈BF₄NO
MW: 479.4013 g/mol
CAS Number: 2270864-45-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR