Ethyl 3-(4-(trifluoromethyl)phenyl)acrylate ≥97%

Supplier: Ambeed
Warning

A1495330-1G A1495330-5G
77624-564EA 24.72 USD
77624-564 77875-617
Ethyl 3-(4-(trifluoromethyl)phenyl)acrylate ≥97%
Ethyl 3-(4-(trifluoromethyl)phenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁F₃O₂
CAS Number: 101466-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR