Remazol brilliant blue R Strength 95-105%, Moisture≤6%

Supplier: Ambeed
Warning

Synonyms: Remazol Brilliant Blue R (C.I. 61200), Sodium 2-((3-((4-amino-9,10-dioxo-3-sulphonato-9,10-dihydroanthracen-1-yl)amino)phenyl)sulphonyl)ethyl sulphate, MFCD00001215, Reactive Blue 19, REACTIVEBLUE19

A1495186-100G A1495186-25G A1495186-500G
77597-696EA 47.15 USD
77597-696 77624-562 77875-597
Remazol brilliant blue R Strength 95-105%, Moisture≤6%
Remazol brilliant blue R

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₂Na₂O₁₁S₃
MW: 626.533 g/mol
MDL Number: MFCD00001215
CAS Number: 2580-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR