Tri(pyridin-3-yl)amine ≥98%

Supplier: Ambeed
Warning

A1494852-1G A1494852-100MG A1494852-250MG
77624-554EA 121.54 USD
77624-554 77875-591 77875-592
Tri(pyridin-3-yl)amine ≥98%
Tri(pyridin-3-yl)amine

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₄
MDL Number: MFCD34469806
CAS Number: 153467-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR