Hexane-1,6-diyl dipropiolate ≥95%

Supplier: Ambeed
Warning

A1494594-100MG A1494594-1G
77875-584EA 63 USD
77875-584 77875-585
Hexane-1,6-diyl dipropiolate ≥95%
Hexane-1,6-diyl dipropiolate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MDL Number: MFCD09039122
CAS Number: 74987-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR