2-Aminobenzene-1,3,5-tricarboxylic acid ≥95%

Supplier: Ambeed
Warning

A1494452-100MG A1494452-250MG
77624-550EA 139.05 USD
77624-550 77875-581
2-Aminobenzene-1,3,5-tricarboxylic acid ≥95%
2-Aminobenzene-1,3,5-tricarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₆
CAS Number: 489-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR