Methyl 5-bromo-6-oxo-1,6-dihydropyridazine-3-carboxylate ≥95%

Supplier: Ambeed
Warning

A1494185-100MG A1494185-1G A1494185-250MG
77875-567EA 101 USD
77875-567 77875-568 77875-569
Methyl 5-bromo-6-oxo-1,6-dihydropyridazine-3-carboxylate ≥95%
Methyl 5-bromo-6-oxo-1,6-dihydropyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₃
MDL Number: MFCD29081496
CAS Number: 1591827-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR