(R)-4-Isopropyl-2-(2-(methylthio)phenyl)-4,5-dihydrooxazole ≥97%
Supplier: Ambeed
|
Warning
|
A1493928-100MG
A1493928-250MG
77624-536EA
87.55
USD
77624-536
77624-538
(R)-4-Isopropyl-2-(2-(methylthio)phenyl)-4,5-dihydrooxazole ≥97%
(R)-4-Isopropyl-2-(2-(methylthio)phenyl)-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NOS MW: 235.3452 g/mol |
CAS Number:
2757083-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...