(S)-2-(2-(Methylthio)phenyl)-4-phenyl-4,5-dihydrooxazole ≥97%

Supplier: Ambeed
Warning

A1493924-250MG A1493924-100MG A1493924-1G
77624-534EA 134.93 USD
77624-534 77875-523 77875-524
(S)-2-(2-(Methylthio)phenyl)-4-phenyl-4,5-dihydrooxazole ≥97%
(S)-2-(2-(Methylthio)phenyl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₅NOS
CAS Number: 154132-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR