2-(2,4,6-Trimethyl-N-(4-((2-(pyrrolidin-1-yl)pyrimidin-4-yl)amino)phenyl)phenylsulfonamido)acetic acid ≥98%

Supplier: Ambeed
Warning

A1493606-10MG A1493606-1MG A1493606-5MG
77597-680EA 64.58 USD
77597-680 77597-682 77597-684
2-(2,4,6-Trimethyl-N-(4-((2-(pyrrolidin-1-yl)pyrimidin-4-yl)amino)phenyl)phenylsulfonamido)acetic acid ≥98%
2-(2,4,6-Trimethyl-N-(4-((2-(pyrrolidin-1-yl)pyrimidin-4-yl)amino)phenyl)phenylsulfonamido)acetic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₉N₅O₄S
MDL Number: MFCD33398564
CAS Number: 2355377-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR