2-(4-Chlorophenyl)naphthalene ≥95%

Supplier: Ambeed
Warning

A1493154-1G A1493154-5G
77875-490EA 11 USD
77875-490 77875-491
2-(4-Chlorophenyl)naphthalene ≥95%
2-(4-Chlorophenyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₆H₁₁Cl
MDL Number: MFCD18415897
CAS Number: 22082-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR