Allyltributylphosphonium bromide ≥95%

Supplier: Ambeed
Warning

A1492881-250MG A1492881-25G
77875-487EA 7 USD
77875-487 77875-488
Allyltributylphosphonium bromide ≥95%
Allyltributylphosphonium bromide

Precaución: for Research Use Only

Formula: C₁₅H₃₂BrP
MDL Number: MFCD01706152
CAS Number: 55894-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR