2-Amino-9-((1S,3R,4R,7S)-7-hydroxy-1-(hydroxymethyl)-2,5-dioxabicyclo[2.2.1]heptan-3-yl)-1,9-dihydro-6H-purin-6-one ≥98%

Supplier: Ambeed

A1491856-100MG A1491856-250MG
77875-472EA 131 USD
77875-472 77875-473
2-Amino-9-((1S,3R,4R,7S)-7-hydroxy-1-(hydroxymethyl)-2,5-dioxabicyclo[2.2.1]heptan-3-yl)-1,9-dihydro-6H-purin-6-one ≥98%
2-Amino-9-((1S,3R,4R,7S)-7-hydroxy-1-(hydroxymethyl)-2,5-dioxabicyclo[2.2.1]heptan-3-yl)-1,9-dihydro-6H-purin-6-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₅O₅
MDL Number: MFCD31707476
CAS Number: 207131-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR