4-Nitrobenzyl (1S,4S)-3-oxo-2-thia-5-azabicyclo[2.2.1]heptane-5-carboxylate ≥98%

Supplier: Ambeed
Warning

A1491357-5G A1491357-100G A1491357-25G
77624-514EA 6.18 USD
77624-514 77875-470 77875-471
4-Nitrobenzyl (1S,4S)-3-oxo-2-thia-5-azabicyclo[2.2.1]heptane-5-carboxylate ≥98%
4-Nitrobenzyl (1S,4S)-3-oxo-2-thia-5-azabicyclo[2.2.1]heptane-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₅S
CAS Number: 151072-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR