Ethyl-6-chloro-5-cyano-2-methylnicotinate ≥95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-chloro-5-cyano-2-methylpyridine-3-carboxylate

A149107-5G A149107-1G
77611-378EA 179.22 USD
77611-378 77671-098
Ethyl-6-chloro-5-cyano-2-methylnicotinate ≥95%
Ethyl-6-chloro-5-cyano-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD00179894
CAS Number: 64119-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR