3-Bromo-2-ethylpyridine 98+%

Supplier: Ambeed
Warning

A1490506-100MG A1490506-250MG A1490506-25G A1490506-5G
77875-456EA 28 USD
77875-456 77875-457 77875-458 77875-459
3-Bromo-2-ethylpyridine 98+%
3-Bromo-2-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MDL Number: MFCD13189503
CAS Number: 38749-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR