3-(Cyanomethoxy)phenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetonitrile

A1490468-250MG A1490468-1G
77624-502EA 39.14 USD
77624-502 77875-450
3-(Cyanomethoxy)phenylboronic acid pinacol ester ≥98%
3-(Cyanomethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
MW: 259.1086 g/mol
MDL Number: MFCD06795674
CAS Number: 936250-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR