Disodium hydrogen citrate ≥98%

Supplier: Ambeed
Warning

A1490379-1000G A1490379-100G A1490379-500G
77592-812EA 44.08 USD
77592-812 77592-814 77592-816
Disodium hydrogen citrate ≥98%
Disodium hydrogen citrate

Precaución: for Research Use Only

Formula: C₆H₆Na₂O₇
MDL Number: MFCD00036367
CAS Number: 144-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR