2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane ≥95%

Supplier: Ambeed
Warning

Synonyms: Tetrabromobisphenol A Bis(2,3-dibromopropyl) Ether, 2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane(Granular)

A1490373-25G A1490373-1000G A1490373-100G A1490373-500G
77624-494EA 11.33 USD
77624-494 77875-444 77875-445 77875-446
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane ≥95%
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane

Precaución: for Research Use Only

Formula: C₂₁H₂₀Br₈O₂
MW: 943.62 g/mol
MDL Number: MFCD00017887
CAS Number: 21850-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR