5'-(4-Formyl-3-methoxyphenyl)-3,3''-dimethoxy-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde ≥98%

Supplier: Ambeed

A1489873-1G A1489873-100MG
77597-648EA 279.83 USD
77597-648 77875-430
5'-(4-Formyl-3-methoxyphenyl)-3,3''-dimethoxy-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde ≥98%
5'-(4-Formyl-3-methoxyphenyl)-3,3''-dimethoxy-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₃₀H₂₄O₆
CAS Number: 2254817-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR