N-(6-((1H-Pyrazol-1-yl)methyl)-4-methoxybenzo[d]isoxazol-3-yl)-2,6-dimethoxybenzenesulfonamide ≥98%

Supplier: Ambeed

Synonyms: PF-9363

A1489779-1MG A1489779-50MG
77597-646EA 23.58 USD
77597-646 77875-425
N-(6-((1H-Pyrazol-1-yl)methyl)-4-methoxybenzo[d]isoxazol-3-yl)-2,6-dimethoxybenzenesulfonamide ≥98%
N-(6-((1H-Pyrazol-1-yl)methyl)-4-methoxybenzo[d]isoxazol-3-yl)-2,6-dimethoxybenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₄O₆S
CAS Number: 2569009-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR