N-((11bR)-2,6-Dimethyl-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide ≥98%

Supplier: Ambeed
Warning

A1489601-100MG A1489601-250MG
77624-466EA 168.92 USD
77624-466 77624-468
N-((11bR)-2,6-Dimethyl-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide ≥98%
N-((11bR)-2,6-Dimethyl-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₂₃H₁₇F₃NO₅PS
MW: 507.4188 g/mol
CAS Number: 2757287-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR