(8-Chlorodibenzo[b,d]furan-1-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1488629-250MG A1488629-5G
77875-411EA 9 USD
77875-411 77875-412
(8-Chlorodibenzo[b,d]furan-1-yl)boronic acid ≥98%
(8-Chlorodibenzo[b,d]furan-1-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₈BClO₃
MW: 246.4541 g/mol
CAS Number: 2173554-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR