1,1'-BIs[(2S,4S)-2,4-bis(1-methylethyl)-1-phosphetanyl]ferrocene ≥97%

Supplier: Ambeed
Warning

A1487884-100MG A1487884-250MG A1487884-50MG A1487884-1G
77597-632EA 176.3 USD
77597-632 77597-634 77597-636 77624-450
1,1'-BIs[(2S,4S)-2,4-bis(1-methylethyl)-1-phosphetanyl]ferrocene ≥97%
1,1'-BIs[(2S,4S)-2,4-bis(1-methylethyl)-1-phosphetanyl]ferrocene

Precaución: for Research Use Only

Formula: C₂₈H₄₄FeP₂
MW: 498.44 g/mol
CAS Number: 224057-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR