1,1'-Bis[(2R,4R)-2,4-dipropyl-1-phosphetanyl]ferrocene stereoisomer ≥97%

Supplier: Ambeed
Warning

A1487883-100MG A1487883-250MG A1487883-50MG A1487883-1G
77597-626EA 176.3 USD
77597-626 77597-628 77597-630 77624-448
1,1'-Bis[(2R,4R)-2,4-dipropyl-1-phosphetanyl]ferrocene stereoisomer ≥97%
1,1'-Bis[(2R,4R)-2,4-dipropyl-1-phosphetanyl]ferrocene stereoisomer

Precaución: for Research Use Only

Formula: C₂₈H₄₄FeP₂
MW: 498.44 g/mol
CAS Number: 268220-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR