1,1'-Bis[(2R,4R)-2,4-dimethyl-1-phosphetanyl]ferrocene stereoisomer ≥97%

Supplier: Ambeed
Warning

A1487881-100MG A1487881-250MG A1487881-50MG A1487881-1G A1487881-5G
77597-616EA 189.63 USD
77597-616 77597-618 77597-620 77624-446 77875-398
1,1'-Bis[(2R,4R)-2,4-dimethyl-1-phosphetanyl]ferrocene stereoisomer ≥97%
1,1'-Bis[(2R,4R)-2,4-dimethyl-1-phosphetanyl]ferrocene stereoisomer

Precaución: for Research Use Only

Formula: C₂₀H₂₈FeP₂
MW: 386.23 g/mol
CAS Number: 268541-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR