8-(4-Nitrophenyl)-1,4-dioxaspiro[4.5]dec-7-ene ≥97%

Supplier: Ambeed
Warning

A1487647-1G A1487647-250MG
77875-389EA 53 USD
77875-389 77875-390
8-(4-Nitrophenyl)-1,4-dioxaspiro[4.5]dec-7-ene ≥97%
8-(4-Nitrophenyl)-1,4-dioxaspiro[4.5]dec-7-ene

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
CAS Number: 2574546-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR