Disodium hexachloroplatinate hexahydrate 98+%

Supplier: Ambeed
Danger

Synonyms: Sodium hexachloroplatinate(IV) hexahydrate, Sodium hexachloroplatinate (IV) hexahydrate

A1487347-5G A1487347-1G A1487347-25G
77624-440EA 202.91 USD
77624-440 77875-382 77875-383
Disodium hexachloroplatinate hexahydrate 98+%
Disodium hexachloroplatinate hexahydrate

Precaución: for Research Use Only

Formula: Cl₆Na₂Pt·6H₂O
MDL Number: MFCD00149172
CAS Number: 19583-77-8
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR