(R)-N-((S)-(2-(Di-tert-butylphosphanyl)phenyl)(naphthalen-2-yl)methyl)-2-methylpropane-2-sulfinamide ≥95%

Supplier: Ambeed
Warning

A1487079-100MG A1487079-250MG A1487079-50MG
77624-434EA 230.72 USD
77624-434 77875-376 77875-377
(R)-N-((S)-(2-(Di-tert-butylphosphanyl)phenyl)(naphthalen-2-yl)methyl)-2-methylpropane-2-sulfinamide ≥95%
(R)-N-((S)-(2-(Di-tert-butylphosphanyl)phenyl)(naphthalen-2-yl)methyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₂₉H₄₀NOPS
MDL Number: MFCD32697222
CAS Number: 2565792-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR