(R)-N-((R)-(3,5-Di-tert-butyl-4-methoxyphenyl)(5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl)methyl)-2-methylpropane-2-sulfinamide ≥95%

Supplier: Ambeed
Warning

A1487059-25MG A1487059-100MG A1487059-50MG
77597-602EA 86.1 USD
77597-602 77624-430 77624-432
(R)-N-((R)-(3,5-Di-tert-butyl-4-methoxyphenyl)(5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl)methyl)-2-methylpropane-2-sulfinamide ≥95%
(R)-N-((R)-(3,5-Di-tert-butyl-4-methoxyphenyl)(5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl)methyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₄₇H₅₆NO₃PS
MW: 745.99 g/mol
MDL Number: MFCD32697188
CAS Number: 2565792-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR